Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008334.1 | NP_001008335.1 | 1 - 2 | 144 | 108 | |
GenBank mRNA | BC085344.1 | - | 1 - 2 | 144 | 7303878 | 108 |
FQ214423.1 | - | 1 - 2 | 186 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |