Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013856.1 | NP_001013878.1 | 10 - 11 | 1310 | 148 | |
GenBank mRNA | BC079308.1 | - | 10 - 11 | 1310 | 7134677 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |