Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199081.1 | NP_954512.1 | 5 - 6 | 610 | 67 | |
GenBank mRNA | AY349135.1 | - | 5 - 6 | 610 | 67 | |
BC085347.1 | - | 5 - 6 | 645 | 7305181 | 67 | |
D87991.1 | - | 5 - 6 | 603 | 67 | ||
FQ233464.1 | - | 5 - 6 | 633 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |