Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014228.1 | NP_001014250.1 | 13 - 14 | 1602 | 88 | |
GenBank mRNA | BC079020.1 | - | 13 - 14 | 1602 | 7113829 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |