Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100158.2 | NP_001093628.1 | 2 - 3 | 306 | 118 | |
GenBank mRNA | BC105875.1 | - | 2 - 3 | 251 | 7130292 | 118 |
CO559861.1 | - | 2 - 3 | 306 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |