Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Havcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100762.1 | NP_001094232.1 | 2 - 3 | 472 | 165 | |
GenBank mRNA | AJ549521.1 | - | 1 - 2 | 340 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |