Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053751.4 | NP_446203.1 | 2 - 3 | 224 | 92 | |
XM_008770244.1 | XP_008768466.1 | 2 - 3 | 591 | 92 | ||
GenBank mRNA | J00801.1 | - | 2 - 3 | 221 | 92 | |
J00802.1 | - | 2 - 3 | 224 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |