Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109412.1 | NP_001102882.1 | 2 - 3 | 1097 | 158 | |
XM_008770361.1 | XP_008768583.1 | 1 - 2 | 1831 | 158 | ||
XM_008770362.1 | XP_008768584.1 | 1 - 2 | 1403 | 158 | ||
GenBank mRNA | BC162020.1 | - | 2 - 3 | 1069 | 9098415 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |