Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053772.2 | NP_446224.1 | 1 - 2 | 111 | 119 | |
GenBank mRNA | FM055544.1 | - | 1 - 2 | 116 | 119 | |
FQ211701.1 | - | 1 - 2 | 141 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |