Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022548.1 | NP_071993.1 | 4 - 5 | 778 | 135 | |
GenBank mRNA | Y13148.1 | - | 4 - 5 | 778 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |