Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106427.1 | NP_001099897.1 | 1 - 2 | 130 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons |