Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019300.1 | NP_062173.1 | 1 - 2 | 70 | 77 | |
GenBank mRNA | FM101110.1 | - | 1 - 2 | 70 | 77 | |
U67914.1 | - | 1 - 2 | 56 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |