Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191703.1 | NP_001178632.1 | 6 - 7 | 1145 | 84 | |
XM_006232225.2 | XP_006232287.1 | 7 - 8 | 1669 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |