Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135873.1 | NP_001129345.1 | 6 - 7 | 732 | 142 | |
XM_006257455.2 | XP_006257517.1 | 6 - 7 | 755 | 142 | ||
XM_008773416.1 | XP_008771638.1 | 6 - 7 | 755 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |