Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191585.1 | NP_001178514.1 | 8 - 9 | 1944 | 120 | |
XM_006257447.2 | XP_006257509.1 | 9 - 10 | 2148 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |