Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053859.2 | NP_446311.1 | 4 - 5 | 664 | 81 | |
GenBank mRNA | EU253553.1 | - | 5 - 6 | 629 | 81 | |
U07609.1 | - | 4 - 5 | 667 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |