Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053599.2 | NP_446051.2 | 4 - 5 | 557 | 111 | |
XM_008761227.1 | XP_008759449.1 | 5 - 6 | 1141 | 111 | ||
XM_008761228.1 | XP_008759450.1 | 4 - 5 | 2089 | 111 | ||
GenBank mRNA | BC070514.1 | - | 4 - 5 | 557 | 7100462 | 111 |
D38056.1 | - | 4 - 5 | 530 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |