Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106437.1 | NP_001099907.1 | 3 - 4 | 817 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |