Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017090.2 | NP_058786.2 | 2 - 3 | 392 | 67 | |
XM_006232518.2 | XP_006232580.1 | 3 - 4 | 984 | 67 | ||
XM_008761107.1 | XP_008759329.1 | 1 - 2 | 1015 | 67 | ||
XM_008761108.1 | XP_008759330.1 | 2 - 3 | 758 | 67 | ||
GenBank mRNA | AB096020.1 | - | 1 - 2 | 255 | 67 | |
BC085746.1 | - | 2 - 3 | 392 | 7190419 | 67 | |
M57405.1 | - | 2 - 3 | 711 | 67 | ||
U60835.1 | - | 2 - 3 | 711 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |