Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198745.2 | NP_942040.1 | 4 - 5 | 374 | 126 | |
GenBank mRNA | BC059155.1 | - | 4 - 5 | 378 | 6887702 | 126 |
FQ213001.1 | - | 4 - 5 | 374 | 126 | ||
FQ213103.1 | - | 4 - 5 | 371 | 126 | ||
FQ213345.1 | - | 4 - 5 | 376 | 126 | ||
FQ213479.1 | - | 4 - 5 | 375 | 126 | ||
FQ213531.1 | - | 4 - 5 | 376 | 126 | ||
FQ213667.1 | - | 4 - 5 | 392 | 126 | ||
FQ213903.1 | - | 4 - 5 | 370 | 126 | ||
FQ214254.1 | - | 4 - 5 | 376 | 126 | ||
FQ214381.1 | - | 4 - 5 | 374 | 126 | ||
FQ214406.1 | - | 4 - 5 | 369 | 126 | ||
FQ214440.1 | - | 4 - 5 | 376 | 126 | ||
FQ214476.1 | - | 4 - 5 | 379 | 126 | ||
FQ214497.1 | - | 4 - 5 | 342 | 126 | ||
FQ214920.1 | - | 4 - 5 | 374 | 126 | ||
FQ216629.1 | - | 4 - 5 | 374 | 126 | ||
FQ224865.1 | - | 4 - 5 | 375 | 126 | ||
FQ226034.1 | - | 4 - 5 | 369 | 126 | ||
FQ230006.1 | - | 4 - 5 | 374 | 126 | ||
FQ230733.1 | - | 4 - 5 | 369 | 126 | ||
FQ233079.1 | - | 4 - 5 | 374 | 126 | ||
FQ233569.1 | - | 4 - 5 | 376 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |