Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037979.1 | NP_001033068.1 | 2 - 3 | 301 | 63 | |
XM_006237183.1 | XP_006237245.1 | 3 - 4 | 607 | 63 | ||
XM_006237184.2 | XP_006237246.1 | 3 - 4 | 478 | 63 | ||
XM_006237185.2 | XP_006237247.1 | 3 - 4 | 493 | 63 | ||
XM_006237186.2 | XP_006237248.1 | 3 - 4 | 622 | 63 | ||
GenBank mRNA | AY724514.1 | - | 3 - 4 | 561 | 63 | |
DQ148392.1 | - | 2 - 3 | 301 | 63 | ||
FQ232508.1 | - | 3 - 4 | 577 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |