Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015005.2 | NP_001015005.2 | 4 - 5 | 460 | 98 | |
XM_006237518.1 | XP_006237580.1 | 5 - 6 | 517 | 98 | ||
GenBank mRNA | BC091696.1 | - | 4 - 5 | 453 | 7301732 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |