Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC092659.1 | - | 3 - 4 | 353 | 116 | |
BC166492.1 | - | 10 - 11 | 1867 | 7110317 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |