Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024153.1 | NP_077067.1 | 8 - 9 | 840 | 59 | |
GenBank mRNA | BC088844.1 | - | 8 - 9 | 847 | 7308634 | 59 |
D63761.1 | - | 8 - 9 | 840 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |