Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138839.2 | NP_620194.1 | 8 - 9 | 901 | 116 | |
XM_006247078.2 | XP_006247140.1 | 8 - 9 | 1379 | 116 | ||
XM_006247079.2 | XP_006247141.1 | 9 - 10 | 923 | 116 | ||
GenBank mRNA | AF411216.1 | - | 8 - 9 | 847 | 116 | |
BC061721.1 | - | 8 - 9 | 901 | 5598561 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |