Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172091.2 | NP_742088.1 | 13 - 14 | 1357 | 58 | |
NM_172092.2 | NP_742089.1 | 13 - 14 | 1365 | 58 | ||
NR_073147.1 | 12 - 13 | 1258 | 58 | |||
XM_006247876.2 | XP_006247938.1 | 13 - 14 | 1471 | 58 | ||
XM_006247877.1 | XP_006247939.1 | 11 - 12 | 1196 | 58 | ||
XM_008768468.1 | XP_008766690.1 | 12 - 13 | 1414 | 58 | ||
GenBank mRNA | BC081700.1 | - | 12 - 13 | 1222 | 7192550 | 58 |
L04796.1 | - | 13 - 14 | 1327 | 58 | ||
M96674.1 | - | 13 - 14 | 1367 | 58 | ||
X68692.1 | - | 13 - 14 | 1327 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |