Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253859.1 | NP_001240788.1 | 15 - 16 | 1420 | 69 | |
GenBank mRNA | BC169112.1 | - | 15 - 16 | 1418 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |