Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108299.1 | NP_001101769.1 | 8 - 9 | 1086 | 115 | |
XM_006247369.2 | XP_006247431.1 | 8 - 9 | 1159 | 115 | ||
XM_006247370.2 | XP_006247432.1 | 8 - 9 | 1107 | 115 | ||
XM_006247371.2 | XP_006247433.1 | 8 - 9 | 1067 | 115 | ||
XM_006247372.2 | XP_006247434.1 | 8 - 9 | 914 | 115 | ||
XM_008768085.1 | XP_008766307.1 | 8 - 9 | 1156 | 115 | ||
XM_008768086.1 | XP_008766308.1 | 8 - 9 | 1142 | 115 | ||
GenBank mRNA | BC168950.1 | - | 8 - 9 | 960 | 9022277 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |