Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012013.1 | NP_001012013.1 | 4 - 5 | 583 | 150 | |
XM_006247508.2 | XP_006247570.1 | 5 - 6 | 571 | 150 | ||
XM_006247509.2 | XP_006247571.1 | 5 - 6 | 573 | 150 | ||
XM_006247510.2 | XP_006247572.1 | 5 - 6 | 572 | 150 | ||
XM_006247511.2 | XP_006247573.1 | 5 - 6 | 571 | 150 | ||
XM_006247512.2 | XP_006247574.1 | 5 - 6 | 572 | 150 | ||
XM_008768284.1 | XP_008766506.1 | 6 - 7 | 674 | 150 | ||
XM_008768285.1 | XP_008766507.1 | 5 - 6 | 562 | 150 | ||
XM_008768286.1 | XP_008766508.1 | 5 - 6 | 573 | 150 | ||
XM_008768287.1 | XP_008766509.1 | 5 - 6 | 572 | 150 | ||
GenBank mRNA | BC076387.1 | - | 4 - 5 | 583 | 7109482 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |