Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134498.2 | NP_001127970.1 | 6 - 7 | 628 | 82 | |
XM_008768343.1 | XP_008766565.1 | 6 - 7 | 2608 | 82 | ||
XM_008768344.1 | XP_008766566.1 | 6 - 7 | 2607 | 82 | ||
XM_008768345.1 | XP_008766567.1 | 6 - 7 | 2606 | 82 | ||
XM_008768346.1 | XP_008766568.1 | 6 - 7 | 2605 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |