Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134498.2 | NP_001127970.1 | 3 - 4 | 412 | 149 | |
XM_008768343.1 | XP_008766565.1 | 3 - 4 | 2392 | 149 | ||
XM_008768344.1 | XP_008766566.1 | 3 - 4 | 2391 | 149 | ||
XM_008768345.1 | XP_008766567.1 | 3 - 4 | 2390 | 149 | ||
XM_008768346.1 | XP_008766568.1 | 3 - 4 | 2389 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |