Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004021.1 | NP_001004021.1 | 7 - 8 | 1247 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |