Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013043.1 | NP_001013061.1 | 4 - 5 | 590 | 133 | |
GenBank mRNA | BC078937.1 | - | 4 - 5 | 590 | 7108345 | 133 |
FQ225276.1 | - | 4 - 5 | 583 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |