Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011892.1 | NP_001011892.1 | 3 - 4 | 147 | 73 | |
XM_006246675.1 | XP_006246737.1 | 3 - 4 | 292 | 73 | ||
GenBank mRNA | BC079362.1 | - | 3 - 4 | 147 | 7109243 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |