Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004201.1 | NP_001004201.1 | 1 - 2 | 244 | 70 | |
XM_006247268.2 | XP_006247330.1 | 1 - 2 | 74 | 70 | ||
GenBank mRNA | BC079258.1 | - | 1 - 2 | 244 | 7133394 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |