Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004201.1 | NP_001004201.1 | 4 - 5 | 506 | 59 | |
XM_006247268.2 | XP_006247330.1 | 4 - 5 | 336 | 59 | ||
GenBank mRNA | BC079258.1 | - | 4 - 5 | 506 | 7133394 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |