Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135039.1 | NP_001128511.1 | 8 - 9 | 1122 | 97 | |
XM_008768044.1 | XP_008766266.1 | 8 - 9 | 1377 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |