Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727774.2 | XP_002727820.1 | 4 - 5 | 596 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |