Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034936.1 | NP_001030108.1 | 2 - 3 | 279 | 91 | |
XM_006246010.2 | XP_006246072.1 | 2 - 3 | 361 | 91 | ||
GenBank mRNA | BC099803.1 | - | 2 - 3 | 279 | 7937864 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |