Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034936.1 | NP_001030108.1 | 9 - 10 | 1124 | 65 | |
XM_006246008.2 | XP_006246070.1 | 10 - 11 | 1246 | 65 | ||
XM_006246009.2 | XP_006246071.1 | 10 - 11 | 1246 | 65 | ||
XM_006246010.2 | XP_006246072.1 | 9 - 10 | 1206 | 65 | ||
XM_006246011.2 | XP_006246073.1 | 10 - 11 | 1058 | 65 | ||
XM_006246012.2 | XP_006246074.1 | 6 - 7 | 2239 | 65 | ||
XM_006246013.2 | XP_006246075.1 | 8 - 9 | 2688 | 65 | ||
XM_006246014.2 | XP_006246076.1 | 8 - 9 | 2589 | 65 | ||
XM_006246015.2 | XP_006246077.1 | 8 - 9 | 2890 | 65 | ||
XM_006246016.2 | XP_006246078.1 | 10 - 11 | 1258 | 65 | ||
XM_006246017.2 | XP_006246079.1 | 7 - 8 | 780 | 65 | ||
GenBank mRNA | BC099803.1 | - | 9 - 10 | 1124 | 7937864 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |