Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133418.1 | NP_596909.1 | 2 - 3 | 246 | 140 | |
GenBank mRNA | AJ223355.1 | - | 2 - 3 | 246 | 140 | |
BC081734.1 | - | 2 - 3 | 264 | 7108552 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |