Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012663.2 | NP_036795.1 | 4 - 5 | 409 | 116 | |
GenBank mRNA | BC074003.1 | - | 4 - 5 | 409 | 7097683 | 116 |
M24105.1 | - | 3 - 4 | 347 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |