Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024256.1 | NP_001019427.1 | 6 - 7 | 651 | 59 | |
GenBank mRNA | BC093402.1 | - | 6 - 7 | 651 | 7389622 | 59 |
FQ231796.1 | - | 4 - 5 | 337 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |