Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770929.1 | XP_008769151.1 | 8 - 9 | 1190 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |