Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diaph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728299.4 | XP_002728345.2 | 19 - 20 | 2331 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |