Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107279.1 | NP_001100749.1 | 2 - 3 | 2760 | 80 | |
XM_006252377.2 | XP_006252439.1 | 1 - 2 | 2576 | 80 | ||
XM_006252378.2 | XP_006252440.1 | 1 - 2 | 2576 | 80 | ||
XM_006252379.1 | XP_006252441.1 | 1 - 2 | 2620 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |