Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024990.2 | NP_001020161.2 | 6 - 7 | 787 | 77 | |
XM_008767565.1 | XP_008765787.1 | 6 - 7 | 878 | 77 | ||
GenBank mRNA | BC091278.1 | - | 6 - 7 | 777 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |