Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053731.2 | NP_446183.2 | 1 - 2 | 1020 | 97 | |
XM_006246567.2 | XP_006246629.1 | 2 - 3 | 1372 | 97 | ||
XM_006246568.2 | XP_006246630.1 | 2 - 3 | 1206 | 97 | ||
GenBank mRNA | AY028417.1 | - | 1 - 2 | 1020 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |