Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053731.2 | NP_446183.2 | 4 - 5 | 1415 | 143 | |
XM_006246567.2 | XP_006246629.1 | 5 - 6 | 1767 | 143 | ||
XM_006246568.2 | XP_006246630.1 | 5 - 6 | 1601 | 143 | ||
GenBank mRNA | AY028417.1 | - | 4 - 5 | 1415 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |