Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148891.2 | NP_683689.1 | 2 - 3 | 621 | 76 | |
GenBank mRNA | AJ492222.1 | - | 2 - 3 | 242 | 76 | |
BC097277.1 | - | 2 - 3 | 621 | 7457194 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |